Learn More
Bullard™ Sentinel™ Faceshield Headgear
Catalog No. 19088776
Change view
Click to view available options
Includes:
Black crown and chin protector
Black crown protector
Product Type:
Sentinel 1 Faceshield Headgear
Sentinel 2 Faceshield Headgear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Product Type
19088776 Black crown and chin protector Sentinel 2 Faceshield Headgear
189993744 Black crown protector Sentinel 1 Faceshield Headgear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19088776 Supplier Bullard™ Supplier No. SENTINEL2
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Does not include faceshield visor (sold separately).
Lightweight molded ABS plastic headgear offers the ultimate value in face protection.
Standard, lightweight face protection with black ABS crown protector. Easily accommodates primary eye protection and facepiece-style respirators. (For ordering information, contact your Fisher Safety Representative).
Key Features
  • Crown strap conforms to shape of head
  • Hand-washable Sportek™ brow pad absorbs moisture for a drier, more comfortable fit
  • Sure-Lock™ ratchet securely adjusts headband from sizes 6 1/2 to 8, or head circumferences from 20 1/2 to 25 in. (52 to 64cm)
Order Info
Note: Does not include faceshield visor (sold separately).
Compliance
Meets ANSI/ISEA Z87.1 standards.
Specifications
Dimensions (L x W) 6.5 to 8 in.
Description Sure-Lock Ratchet
Product Type Sentinel 2 Faceshield Headgear
For Use With (Equipment) For use with Bullard 840MX2 (acetate), 840PX2 (polycarbonate), 840PX2W3 (Shade 3 polycarbonate), 840PX2W5 (Shade 5 polycarbonate) or 840TX2 (Tritan) visors. Visor sold separately.
Includes Black crown and chin protector
Lens Material ABS