Learn More
Bullard™ HC2400 Hot/Cool Climate Control Tube
Catalog No. 19087970
Change view
Click to view available options
Includes:
Flow Control Valve, Belt Bracket, Belt and Heat Shield
Catalog No. Includes
19087970 Flow Control Valve, Belt Bracket, Belt and Heat Shield
1 options
Catalog No. 19087970 Supplier Bullard™ Supplier No. HC2400
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
NIOSH-approved accessory for use with certain SAR systems to supply a continuous flow or warm or cool air.
  • Includes:
    • Hot/Cold Tube
    • Flow Control Valve
    • Belt Bracket
    • Belt and Heat Shield
Specifications
Certifications/Compliance NIOSH
For Use With (Equipment) Series CC20 Airline Respirator Hoods
Includes Flow Control Valve, Belt Bracket, Belt and Heat Shield
Model HC2400
Product Type Hot/Cold Tube