Learn More
Bullard™ Constant Flow Assemblies for RT Series Airline Respirators
Description
- Constant flow accessory
- For use with RT series airline respirator
- Flow control assemblies
- Nipple types include bayonet, CEJN, industrial interchange, schrader, or snap-tite
- Does not include breathing tube
Specifications
Specifications
| Connection Type | 0.25 in. Snap-Tite Steel Nipple |
| Controller Type | Constant Flow |
| Material | Stainless Steel |
| Product Type | Flow Control Assembly |
| For Use With (Equipment) | RT Series Airline Respirator |
| Model | F32 |