Learn More
Bullard™ Constant Flow Assemblies for RT Series Airline Respirators
Catalog No. 361003804
Change view
Click to view available options
Connection Type:
0.25 in. Industrial Interchange (Hansen-compatible) Nipple
0.25 in. Snap-Tite Steel Nipple
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
361003804 0.25 in. Industrial Interchange (Hansen-compatible) Nipple
361003805 0.25 in. Snap-Tite Steel Nipple
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361003804 Supplier Bullard™ Supplier No. F30
Only null left
Flow control assembly for use with RT Series respirator, does not include breathing tube.
  • Constant flow accessory
  • For use with RT series airline respirator
  • Flow control assemblies
  • Nipple types include bayonet, CEJN, industrial interchange, schrader, or snap-tite
  • Does not include breathing tube
Specifications
Connection Type 0.25 in. Industrial Interchange (Hansen-compatible) Nipple
Product Type Flow Control Assembly
For Use With (Equipment) CC20 Series Respirators
Model F30