missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ Faceshield Visors

Catalog No. p-94124
Change view
Click to view available options
Lens Material:
Acetate
Dimensions (L x W):
7 x 15 in.
10 x 15 in.
Thickness (English):
0.040 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Material Dimensions (L x W) Thickness (English)
19021321 Acetate 10 x 15 in. 0.040 in.
19088776A Acetate 7 x 15 in. 0.040 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19021321 Supplier Bullard™ Supplier No. 1040
Only null left

For use with Bullard headgear

  • Clear visors accommodate safety spectacles, goggles, and respirators to protect face from numerous industrial hazards
  • Flat acetate visors are for chemical applications and light impact protection
  • Heat-formed polycarbonate visors are for high-heat operations with little or no chemical exposure, and are curved for easy attachment and reduced stress cracking
  • Visors are 15 in. wide (20.3cm) and available in 6, 8, and 10 in. lengths (15.2, 20.3, 25.4cm)

Order Info

Do not use any faceshield unless suitable primary eye protection is used.

Compliance

Meets ANSI/ISEA Z87.1 and ANSI Z87 standards.

Specifications

Dimensions (L x W) 10 x 15 in.
Shape Flat
Thickness (English) 0.040 in.
Thickness (Metric) 1.0 cm
Width (English) 15 in.
Width (Metric) 38.1 cm
Certifications/Compliance ANSI Z87.1
Lens Tint Clear
Product Type Clear flat acetate visor
Length (English) 8 in.
Length (Metric) 20.3 cm
Lens Material Acetate
Show More Show Less