missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bullard™ Loose Fitting Facepieces for PA20, PA30, PA30IS, EVA, EVAHL PAPRs
Loose fitting facepieces provide comfortable and effective protection from a wide range of respiratory hazards.
$634.76 - $680.77
Specifications
| Assigned Protection Factor (APF) | 25 |
|---|---|
| Product Type | Loose Fitting Facepiece |
| For Use With (Application) | Pharmaceutical manufacturing, general industry and maintenance, woodworking, spray foam, painting, healthcare |
| For Use With (Equipment) | Pa20, pa30, pa30is, eva™, eva™hl powered air purifying respirator |
| Latex Free | Yes |
| Catalog Number | Mfr. No. | Includes | Size | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Size | Price | Quantity | ||||
|
19-824-430
|
Bullard™
20LF2S |
Narrow profile, green twist lock connection | Small |
Case of 10 Each for $634.76
|
|
||||
|
18-998-003
|
Bullard™
20LF2M |
Narrow profile, blue twist lock connection | Medium |
Case of 10 Each for $680.77
|
|
||||
|
29-635-9915
|
Bullard™
20LF2L |
Narrow profile, black quick-attach twist lock connection | Large |
Pack of 10 Each for $653.48
|
|
||||
|
29-635-9188
|
Bullard™
20LFM |
Traditional fit, Quick-attach twist lock connection | Medium |
Pack of 10 Each for $634.76
|
|
||||
|
19-021-233C
|
Bullard™
20LFL |
Traditional fit, Quick-attach twist lock connection |
Case of 10 Each for $661.10
|
|
|||||
Description
- Loose fitting facepiece
- Head covers made of Tychem
- Sewn-in headband suspension
- Twist-lock breathing tube connection
- For use with PAPR and SAR systems
- Choose from a variety of sizes and color connections
- Acetate lens material
- NIOSH approved
Specifications
| 25 | |
| Pharmaceutical manufacturing, general industry and maintenance, woodworking, spray foam, painting, healthcare | |
| Yes |
| Loose Fitting Facepiece | |
| Pa20, pa30, pa30is, eva™, eva™hl powered air purifying respirator |