Learn More
Bullard™ Evolutionary Air Hazardous Location (EVAHL) PAPR System Accessory, QC Hood
Catalog No. p-4702228
Change view
Click to view available options
Product Type:
Double Bib Hood
Single Bib Hood
Includes:
Clamp connector style/threaded connector style
Vinyl decon belt, lamp connector style/threaded connector style
For Use With (Application):
Ideal for painting, chemical handling, healthcare, pharmaceutical
Painting, chemical handling
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes For Use With (Application)
17989519 Double Bib Hood Vinyl decon belt, lamp connector style/threaded connector style Ideal for painting, chemical handling, healthcare, pharmaceutical
17989518 Double Bib Hood Clamp connector style/threaded connector style Painting, chemical handling
17989520 Single Bib Hood Clamp connector style/threaded connector style Ideal for painting, chemical handling, healthcare, pharmaceutical
17989521 Single Bib Hood Clamp connector style/threaded connector style Ideal for painting, chemical handling, healthcare, pharmaceutical
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17989519 Supplier Bullard™ Supplier No. EVAHL20TICTDB
Only null left
Designed to be used with the Evolutionary Air Hazardous Location Powered Air Purifying Respirator
  • Dupont™ Tychem QC hood
  • Double bib with ratchet suspension
  • Sealed seams (Cat. No. 17989516 only)
  • Threaded connector style
Certifications
ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Specifications
Battery Type Lithium-ion Polymer
Certifications/Compliance ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Product Type Double Bib Hood
For Use With (Application) Ideal for painting, chemical handling, healthcare, pharmaceutical
Includes Vinyl decon belt, lamp connector style/threaded connector style
Suspension Type Ratchet suspension