Learn More
Bullard™ Hood Battery for EVAHL PAPR System
Catalog No. 17989541
Catalog No. 17989541 Supplier Bullard™ Supplier No. EVAHLBAT1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed to be used with the Evolutionary Air Hazardous Location Powered Air Purifying Respirator.
  • Black
Compliance
Meets NIOSH; ANSI™/ISA 2.12.01-2007 and CSA approvals for Class I, Division 2, Groups A, B, C, D, T6; Class II, Division 2 Groups F, G; Class III.
Specifications
Product Type Battery Pack
Model EVAHLBAT1