Learn More
Bullard™ Evolutionary Air Hazardous Location (EVAHL) PAPR System Accessory, Double Bib Hood
Catalog No. p-4702222
Change view
Click to view available options
Product Type:
Double Bib Hood
Double Bib Saranex™ Hood
Double Bib Suspension-free Hood
Double Bib Suspension-free Hood with Vinyl Decon Belt
For Use With (Application):
Ideal for painting, chemical handling, healthcare, pharmaceutical
Painting, chemical handling
Includes:
Ratchet suspension
S51 hard hat, ratchet suspension, blower, belt, battery, breathing tube, headtop & charger
Saranex™ Hood, Ratchet suspension
Suspension Type:
Ratchet suspension
Suspension-free
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Application) Includes Suspension Type
17989510 Double Bib Hood Painting, chemical handling Ratchet suspension Ratchet suspension
17989512 Double Bib Saranex™ Hood - Saranex™ Hood, Ratchet suspension Ratchet suspension
17989514 Double Bib Hood - S51 hard hat, ratchet suspension, blower, belt, battery, breathing tube, headtop & charger Ratchet suspension
17989531 Double Bib Suspension-free Hood with Vinyl Decon Belt Ideal for painting, chemical handling, healthcare, pharmaceutical - -
17989530 Double Bib Suspension-free Hood - - Suspension-free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17989510 Supplier Bullard™ Supplier No. EVAHL20SICHT
Only null left
Designed to be used with the EVAHL Powered Air Purifying Respirator
  • Ideal for painting, chemical handling, healthcare, pharmaceutical applications
  • Choice of hood configurations
  • Airflow rate: 8.5/7 cfm
  • 77dBA low voltage and 77dBA low flow alarm
  • Lithium ion polymer battery (1-year life span)
Certifications
ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Specifications
Battery Type Lithium-ion Polymer
Certifications/Compliance ISO 9001, NIOSH, CSA, Class I Division 2, Class II Division 2, Class III
Product Type Double Bib Hood
For Use With (Application) Painting, chemical handling
Includes Ratchet suspension
Suspension Type Ratchet suspension