missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ Blower Assembly for EVAHL Powered Air Purifying Respirator

Catalog No. 17989534
Change view
Click to view available options
Battery Life:
4 to 12 hours
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Battery Life
17989534 4 to 12 hours
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17989534 Supplier Bullard™ Supplier No. EVAHL1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

EVAHL blower is approved for use in hazardous locations and designed to complete the EVAHL powered air purifying respirator when assembled with a hood, breathing tube, and filter cartridge.

  • EVAHL PAPR blower assembly designed for hazardous locations
  • Choice of assembly configurations 
  • NIOSH approved EVAHL powered air purifying system consists of four parts:
    • Hood
    • Breathing Tube
    • Blower assembly
    • Filter cartridge
  • User controlled, choose from high or low speed
  • Brushless motor technology

Compliance

NIOSH; ANSI™/ISA 2.12.01-2007 and CSA approvals for Class I, Division 2, Groups A, B, C, D, T6; Class II, Division 2 Groups F, G; Class III.

Specifications

Battery Life 4 to 12 hours
Battery Type Rechargeable Lithium Polymer
Color Black, Blue
Product Type Motor Blower
Flow Rate 7.0 to 8.5 CFM
For Use With (Application) Hazardous locations, pharmaceutical manufacturing, healthcare, laboratories, painting, powder coating, general industries
For Use With (Equipment) EVAHL PAPR Systems
Model EVAHL1