missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ EVAHL PAPR System For Use with Hazardous Locations

Catalog No. p-4917935
Change view
Click to view available options
Size:
Large (Narrow)
Medium (Narrow)
Medium
Belt Material:
Foam
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Belt Material
17989501 Medium Foam
17989500 - Foam
17989503 Medium (Narrow) Foam
17989502 Large (Narrow) Foam
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17989501 Supplier Bullard™ Supplier No. EVAHL20LFM
Only null left

Narrow profile PAPR offers workers protection in hazardous locations. System includes loose-fitting facepiece, blower, belt, battery, breathing tube, charger, air flow indicator, and head top with ratchet suspension.

  • HazLoc rated†
  • For Class I, Division 2, Groups A, B, C, D T6 (Gases and Vapors)
  • Class II, Division 2, Groups F, G (Dusts)
  • Class III (Fibers or Flyings)
  • Simple, one button operation
  • Choice of two air speeds
  • Single filter design
  • Active Flow Technology responds to air flow conditions
  • 25 Assigned Protection Factor (APF) for Loose Fitting Headtops
  • Independent, 3rd Party Documentation for Assigned Protection Factor of 1000 for certain style Hoods (RT, CC20) and Spectrum Full face mask
  • Dual alarms for low flow and low battery
  • Visual fuel gauge
  • 10X the life of most brush motors
  • Genius Charger compatible
†Not for use in Oxygen deficient atmospheres

Compliance

Meets NIOSH; ANSI™/ISA 2.12.01-2007 and CSA approvals for Class I, Division 2, Groups A, B, C, D, T6; Class II, Division 2 Groups F, G; Class III.

†CSA International Certificate 2510928 per ANSI/ISA 12.12.01-2007

Specifications

ANSI Standard ANSI/ISA 2.12.01-2007
Battery Life 7 to 8 hours
Battery Type Lithium-ion Polymer
Belt Material Foam
Certifications/Compliance NIOSH
Certifications (CSA) CSA Class I, Division 2, Groups A, B, C, D, T6, Class II, Division 2 Groups F, G, Class III
Product Type PAPR System with Facepiece
Size Medium
Flow Rate 8.5/7 CFM
For Use With (Application) Painting, chemical handling, healthcare, pharmaceutical
Model EVAHL20LFM
Show More Show Less