missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ EVA™ PAPR Breathing Tube Covers

Catalog No. 19824424
Change view
Click to view available options
Material:
Polypropylene
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Color
19824424 Polypropylene Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19824424 Supplier Bullard™ Supplier No. POLYBTC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disposable breathing tube covers keep the EVA™ PAPR clean from debris.

Specifications

Color Clear
Material Polypropylene
Product Type Breathing Tube Cover
For Use With (Equipment) EVA™ PAPR Systems
Model POLYBTC
Length (English) 456 in.
Length (Metric) 1158.2 cm