missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bullard™ EVA™ Powered-Air Purifying Respirator Shower Caps
Shower cap accessories and replacement parts for EVA™ PAPR systems.
$63.50 - $63.50
Specifications
| Color | White |
|---|---|
| Disposable | Reusable |
| Product Type | Shower Cap |
| For Use With (Application) | For Decon Shower |
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | |||||
|
17-986-283
|
Bullard™
PAPRSC2 |
EVA™ FC3 PAPR System |
N/A
|
|
|||||
Description
- Parts for EVA™ PAPR unit
- Shower cap accessories
Specifications
| White | |
| Reusable | |
| Shower Cap | |
| For Decon Shower |