missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ Chargers for EVA™ Powered-Air Purifying Respirators

Catalog No. p-4898718
Change view
Click to view available options
No. of Ports:
One
Six
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Ports
17986279 One
17986281 Six
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17986279 Supplier Bullard™ Supplier No. EVASMC
Only null left

Chargers designed to be used with the Evolutionary Air (EVA) Powered Air-Purifying Respirator (PAPR) system.

  • Choice of single or six port configurations to suit most needs

Specifications

Color Black
Product Type Single-Unit Charging Station
For Use With (Equipment) EVA™ PAPR Systems
Includes Fuel gauge
Model EVASMC
No. of Ports One