missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ HEPA Filter and Combination Cartridges for EVA™ Powered Air Purifying Respirator

Catalog No. 17986282
Change view
Click to view available options
Product Type:
HEPA Filter Cartridge
HEPA/Chemical Cartridge
HEPA/OV/Chemical Cartridge
Color:
Magenta
Magenta/Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color
17986282 HEPA Filter Cartridge Magenta
19824560 HEPA/OV/Chemical Cartridge Magenta/Yellow
19808700 HEPA/Chemical Cartridge -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17986282 Supplier Bullard™ Supplier No. PAPRFC3
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Filters and combination cartridges are designed for use with EVA series PAPR systems and provide protection against particulates.

  • Protection: Particulates, Ammonia, Formaldehyde, Methylamine, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, and Hydrogen Fluoride
  • Particulate Efficiency: High Efficiency Particulate Absolute (HEPA); 99.99% vs. a mass mean 0.3 micron aerosol at 85 LPM; Media secured with urethane glue around perimeter
  • Filter Housing Material: ABS Plastic
  • Sorbent Bed: Activated Carbon (Chromium Free)
  • Regulatory Approvals: USA; NIOSH 42 CFR Part 84; TC-23C-2886 (EVA PAPR with Loose Fitting Headtops) - per test procedures: TEB-APR-STP-0001, RCT-APR-STP-0035, RCT-APR-STP-0036, RCT-APR-STP-0040, RCT-APR-STP-0042, RCT-APR-STP-0046, RCT-APR-STP-0048
  • Packaging: 6 per box; Each filter individually sealed in plastic
  • Lot Code Identification: Yes
  • Expiration/Shelf Life: 5 years (unopened with proper storage conditions)

Compliance

Tested in accordance to NIOSH 42 CFR Part 84; EN 12941; CE 1094 (EVA PAPR with Loose Fitting Headtops only) requirements.

Specifications

Certifications/Compliance NIOSH
Connection Type Threaded
Color Magenta
Product Type HEPA Filter Cartridge
For Use With (Equipment) EVA™ PAPR Systems