missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ Belts for EVA™ Powered-Air Purifying Respirators

Catalog No. 17986284
Change view
Click to view available options
Material:
Foam
Vinyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
17986284 Foam
17989618 Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17986284 Supplier Bullard™ Supplier No. EVABELT1
Only null left

Accessories and replacement parts for EVA™ PAPR systems.

  • Parts for EVA™ PAPR unit
  • Choose from vinyl or foam material
  • Can be paired with extenders

Specifications

Color Black
Disposable Reusable
Material Foam
Product Type Belt
For Use With (Equipment) EVA™ PAPR Systems
Model EVABELT1