missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ EVA™ Powered Air-Purifying Respirator Battery Packs

Catalog No. p-4898717
Change view
Click to view available options
Color:
Black
Green
For Use With (Equipment):
EVA™ PAPR System Hoods
EVA™ PAPR System Masks
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color For Use With (Equipment)
17986277 Black EVA™ PAPR System Hoods
17986278 Green EVA™ PAPR System Masks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17986277 Supplier Bullard™ Supplier No. EVABAT1
Only null left

Batteries provide power to EVA™ PAPR system blower units.

  • Battery is for use with EVA PAPR hoods (black) or loose-fitting facepieces (green)
  • Lithium
  • Rechargeable

Specifications

Battery Life 7 to 8 hr.
Battery Type Lithium
Color Black
Product Type Battery Pack
For Use With (Equipment) EVA™ PAPR System Hoods
Includes Fuel gauge
Model EVABAT1