Learn More
Bullard™ EVA PAPR System with Loose-Fitting Hood
Catalog No. 17986261
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17986261 Medium
17986260 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17986261 Supplier Bullard™ Supplier No. EVA20LF2M
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Evolutionary Air (EVA) Powered Air-Purifying Respirator (PAPR) uses a battery-powered fan to draw in ambient air through a filter cartridge then delivers filtered air to the hood. System includes PAPR hood system, EVA blower, battery, charger, and breathing tube.
  • Uniquely designed, streamlined form makes PAPR lightweight, eliminates bulky corners that can dig into the wearer's back, and balances the unit's weight for maximum comfort
  • Easy-to-read fuel gauge displays remaining battery capacity at a glance
  • Battery status can be checked at any time during use
  • Bullard Active Flow Technology automatically responds to a worker's need for more or less air flow
  • Two distinct audible alarms notify wearers of low battery and low air flow
  • Alarms are designed so the sound of the alarm travels through the breathing tube directly to the wearer's ear
  • Powered by a long-lasting 10,000-hour brushless motor
  • Single filter design for easy use
  • Filter locking tab notifies the user that the filter is in place with both a visual indication and audible click
Order Info
A NIOSH approved EVA PAPR system consists of the following properly-assembled components: (1) hood or mask; (2) breathing tube; (3) blower assembly; (4) purifying filter cartridge. Check model sytem components carefully before ordering.
Compliance
NIOSH
Specifications
Assigned Protection Factor (APF) 1000
Battery Life 7 to 8 hours
Belt Material Foam
Product Type PAPR System with Hood
Size Medium
Facepiece/Headtop Model Hood
For Use With (Application) Pharmaceutical manufacturing, healthcare, painting, coatings, grinding, welding
For Use With (Equipment) CC20 Series and RT Series Hoods, GR50 Series Nomex grinding hood, FAMB2 Series half mask, and Spectrum Series Full Facepiece
Includes Powered air purifying respirator system, eva™ blower, battery, charger, breathing tube and hood/facepiece
Model EVA20LF2M