Learn More
Bullard™ Clean Air Box (CAB) Series Fixed Breathing Air Filtration/CO Monitoring, Two to Four Worker System
Catalog No. 19100655
Change view
Click to view available options
Product Type:
Manifold Assembly
Connection Type:
Hansen outlet couplings
Schrader outlet couplings
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Connection Type
19100655 Manifold Assembly Schrader outlet couplings
19100654 Manifold Assembly Hansen outlet couplings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
These units will not remove Carbon Monoxide (CO), Carbon Dioxide (CO2), Nitrogen, or other toxic gases or fumes. These units will not increase the oxygen content of an air supply and should not be used when air entering the system is oxygen deficient.
Catalog No. 19100655 Supplier Bullard™ Supplier No. CAB50PMSC
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These units will not remove Carbon Monoxide (CO), Carbon Dioxide (CO2), Nitrogen, or other toxic gases or fumes. These units will not increase the oxygen content of an air supply and should not be used when air entering the system is oxygen deficient.
Wall-mount clean air boxes offer 3-stage filtration with carbon monoxide (CO) monitoring to comply with OSHA’s requirement for providing workers with Grade D breathing air.
  • Clean Air Boxes (CAB) wall units made of stainless steel
  • 50 cfm with four outlets
  • Offer 3-stage filtration with carbon monoxide (CO) monitoring
  • Provide workers with Grade D breathing air
  • Three stages of filtration remove oil, water, particulate, oil vapor and odors
  • Designed for multiple users with or without independent regulators in a portable or fixed option
Specifications
Color Yellow
Connection Type Schrader outlet couplings
Flow Rate 50 cfm
Material Stainless Steel
Includes 110 V Power Cord
Model CAB50PMSC
Product Type Manifold Assembly