Learn More
Bullard™ Spectrum Full Face Airline Respirator System
Catalog No. 19824318
Change view
Click to view available options
Size:
One Size
Catalog No. Size
19824318 One Size
1 options
Catalog No. 19824318 Supplier Bullard™ Supplier No. SPECLSYS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SAR system includes head strap, regulator, lens, padded shoulders and is ideal for spray painting applications, chemical handling, and pharmaceutical manufacturing.
Specifications
Product Type Supplied Air Respirator System
Size One Size
For Use With (Equipment) Spectrum series airline respirator system
Includes Spec35L respirator assembly, EDP10 free-air™ pump, V20100ST hose, LC lens covers
Model SPECLSYS