missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl Bulk Shelving Shelf Divider
Description
- Shelf divider accessory
- For use with bulk shelving system
Specifications
Specifications
| Width (English) | 24 in. |
| Includes | Divider |
| Color | Chrome |
| For Use With (Application) | Shelving |
| Material | Steel |