missing translation for 'onlineSavingsMsg'
Learn More

Wanzl Bulk Shelving Middle Cross Bar

Catalog No. p-7225011
Change view
Click to view available options
Width (English):
36 in.
48 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
11222156 36 in.
11222157 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11222156 Supplier Wanzl Supplier No. T.BCR36CB
Only null left

Middle Cross Bar for use with bulk shelving.

  • Middle Cross Bar
  • Replacement part for bulk shelving system

Specifications

Width (English) 36 in.
Color Chrome
For Use With (Application) Shelving
Includes 1 Middle Cross Bar
Material Steel