missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl Bulk Shelving Middle Cross Bar
Description
- Middle Cross Bar
- Replacement part for bulk shelving system
Specifications
Specifications
| Width (English) | 48 in. |
| Includes | 1 Middle Cross Bar |
| Color | Chrome |
| For Use With (Application) | Shelving |
| Material | Steel |