missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl Bulk Shelving CF Back Label Holder
Description
- CF Back Label Holder
- Color: Black
- Accessory for bulk shelving system
Specifications
Specifications
| Color | Black |
| For Use With (Application) | Shelving |
| Includes | Label Holder |
| Width (English) | 36 in. |
| Material | Steel |