missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl Bulk Shelving Boxed Steel Frame
Description
- Boxed Steel Frame
- Replacement part for bulk shelving system
Specifications
Specifications
| Color | Chrome |
| Depth (English) | 24 in. |
| For Use With (Application) | Shelving |
| Includes | 1 Boxed Steel Frame |
| Height (English) | 60 in. |
| Material | Steel |