Learn More
Poltex Bulk Loose PPE Dispenser
Catalog No. 22000319
Encompass
Change view
Click to view available options
Mounting:
Keyhole Slots
Magnetic Mount
VHB Tape
Description:
Holds and dispenses bulk PPE
Holds and dispenses two types of bulk PPE
Dimensions (D x W x H) Interior:
10 x 10 x 4 in.
15 x 15 x 4 in.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Description Dimensions (D x W x H) Interior
22000319 VHB Tape Holds and dispenses two types of bulk PPE 15 x 15 x 4 in.
22000311 Keyhole Slots Holds and dispenses bulk PPE 10 x 10 x 4 in.
22000316 VHB Tape Holds and dispenses two types of bulk PPE 10 x 10 x 4 in.
22000315 Magnetic Mount Holds and dispenses two types of bulk PPE 10 x 10 x 4 in.
22000317 Keyhole Slots Holds and dispenses two types of bulk PPE 15 x 15 x 4 in.
22000314 Keyhole Slots Holds and dispenses two types of bulk PPE 10 x 10 x 4 in.
22000312 Magnetic Mount Holds and dispenses bulk PPE 10 x 10 x 4 in.
22000313 VHB Tape Holds and dispenses bulk PPE 10 x 10 x 4 in.
22000318 Magnetic Mount Holds and dispenses two types of bulk PPE 15 x 15 x 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 22000319 Supplier Poltex Supplier No. SPBLKPPE15T
Get a Poltex Pipette Holder with $1,500 Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Holds and dispenses PPE including masks, gowns, beard covers, hair nets, bouffants, booties, etc.
Sliding lid helps prevent dust accumulation
Specifications
Dimensions (D x W x H) Interior 15 x 15 x 4 in.
Material PETG
Mounting VHB Tape
Product Style Sliding Lid
Description Holds and dispenses two types of bulk PPE