missing translation for 'onlineSavingsMsg'
Learn More

Poltex Bulk Eyewear Dispenser with Magnets

Catalog No. 22000268
Encompass
Change view
Click to view available options
Bulk Eyewear Dispenser with Magnets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22000268 Bulk Eyewear Dispenser with Magnets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22000268 Supplier Poltex Supplier No. BULKEYEM
Get a Poltex Pipette Holder with $1,500 Purchase
View all promotions

Poltex Bulk Eyewear Dispenser loads from the top, dispenses from the bottom

  • Holds and dispenses protective eyewear
  • Durable PETG construction
  • Powerful magnets for mounting

Specifications

Width (English) 8 in.
Height (English) 15.5 in.
Color Clear
Depth (English) 4.5 in.
Product Type Eyewear Dispenser
No. per Pack 1 Ea.