missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Chemical Identifiers
| Incandescent |
Specifications
| DC Bayonet (BA15D) | |
| 100 hr. | |
| 50 w | |
| Base anode: SFA6-2 (Contains Mercury!), BA15D/DC Bayonet | |
| 8.5 mm | |
| 2.08 in. | |
| 53 mm | |
| 2000 lm |
| Incandescent | |
| 6 V | |
| Clear | |
| 0.33 in. | |
| Microscope | |
| 0.86 in. | |
| 2.2 cm |