missing translation for 'onlineSavingsMsg'
Learn More

Bulbtronics™ Halogen Light Bulbs

Catalog No. 14395902
Encompass
Change view
Click to view available options
Voltage:
220/240 V
6 V
12 V
23 V
24 V
Wattage:
10 W
20 W
35 W
50 W
100 W
250 W
Base Style:
2 Pin (G4)
2 Pin (G6.35)
2 Pin (GX5.3)
2 Pin (GY6.35)
2 Pin (GZ6.35)
E11 Mini Candelabra
PY16-1.25
Shape:
MR16
T2.75
T3
T3.5
T4
T9
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage Wattage Base Style Shape
14395902 220/240 V 250 W E11 Mini Candelabra T4
14395870 12 V 100 W 2 Pin (GY6.35) T3.5
14395869 12 V 100 W 2 Pin (GY6.35) T4
14395871 24 V 250 W 2 Pin (GX5.3) MR16
14395873 6 V 20 W 2 Pin (G4) T2.75
14395874 12 V 100 W 2 Pin (GZ6.35) MR16
14395892 12 V 35 W PY16-1.25 T9
14395893 23 V 50 W 2 Pin (G6.35) T4
14395897 6 V 10 W 2 Pin (G4) T3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Catalog No. 14395902 Supplier Bulbtronics™ Supplier No. 0079208

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Specifications

Base Style E11 Mini Candelabra
Light Source Halogen
Voltage 220/240 V
Wattage 250 W
Color Clear
Diameter (English) 0.55 in.
Diameter (Metric) 1.39 cm
Length (English) 3.34 in.
Length (Metric) 8.48 cm
Shape T4
Product Type Halogen Bulb
Show More Show Less