missing translation for 'onlineSavingsMsg'
Learn More

Bulbtronics™ Halogen Light Bulbs

Supplier:  Bulbtronics™ 0049776

Encompass

 View more versions of this product

Catalog No. 14395897


Only null left

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


Description

Description

Specifications

Chemical Identifiers

Halogen
Halogen Bulb

Specifications

2 Pin (G4)
300 hr.
10 W
0.37 in.
Projector
0.94 in.
2.38 cm
T3
Halogen
6 V
Clear
0.95 cm
1.48 in.
3.75 cm
150 lm
Halogen Bulb
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions