missing translation for 'onlineSavingsMsg'
Learn More

Bulbtronics™ Halogen Light Bulbs

Catalog No. 14395870
Encompass
Change view
Click to view available options
Voltage:
220/240 V
6 V
12 V
23 V
24 V
Wattage:
10 W
20 W
35 W
50 W
100 W
250 W
Base Style:
2 Pin (G4)
2 Pin (G6.35)
2 Pin (GX5.3)
2 Pin (GY6.35)
2 Pin (GZ6.35)
E11 Mini Candelabra
PY16-1.25
Shape:
MR16
T2.75
T3
T3.5
T4
T9
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage Wattage Base Style Shape
14395870 12 V 100 W 2 Pin (GY6.35) T3.5
14395869 12 V 100 W 2 Pin (GY6.35) T4
14395871 24 V 250 W 2 Pin (GX5.3) MR16
14395873 6 V 20 W 2 Pin (G4) T2.75
14395874 12 V 100 W 2 Pin (GZ6.35) MR16
14395892 12 V 35 W PY16-1.25 T9
14395893 23 V 50 W 2 Pin (G6.35) T4
14395897 6 V 10 W 2 Pin (G4) T3
14395902 220/240 V 250 W E11 Mini Candelabra T4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Catalog No. 14395870 Supplier Bulbtronics™ Supplier No. 0000926

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Specifications

Base Style 2 Pin (GY6.35)
Light Source Halogen
Lighting Life 50 hr.
Voltage 12 V
Wattage 100 W
Color Clear
Diameter (English) 0.45 in.
Diameter (Metric) 1.14 cm
For Use With (Equipment) Microscope, collimator
Length (English) 1.73 in.
Length (English) Light Centre 1.18 in.
Length (Metric) 4.39 cm
Length (Metric) Light Centre 2.99 cm
Lumen 3600 lm
Shape T3.5
Product Type Halogen Bulb
Show More Show Less