missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Bug Viewer with Magnifier Lid
Designed for observing bugs, insects, flowers, and more!
Supplier: Eisco™ BI0270
Description
- Optically polished glass lens
- Molded in tough ABS plastic
- Designed for viewing bugs, insects, flowers and more
- Grid screen printed on bottom of jar to easily measure bugs or specimens
- Lid can be used as separate magnifier
- 4X Magnifying Lens: 1.38 in. (35 mm)
Specifications
| Early Childhood, Elementary, Intermediate - Middle School, High School, Higher Ed | |
| 4 to 20 | |
| Viewing Bugs, Insects, Flowers and More | |
| 40 mm (Jar) | |
| 45 mm (Jar) |
| Pre-K to Undergraduate | |
| 1.57 in. (Jar) | |
| ABS Plastic | |
| 1.77 in. (Jar) | |
| Magnifier |