missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Water, Waste Reservoir

Catalog No. 6280163
Encompass
Change view
Click to view available options
Quantity:
4 Reservoirs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6280163 4 Reservoirs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6280163 Supplier Thermo Fisher Scientific Supplier No. 6280163

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Type Waste Reservoir
For Use With (Equipment) 3100 Genetic Analyzer
Quantity 4 Reservoirs
Content And Storage Store at room temperature
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.