missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate Buffer, pH 7, for Residual Chlorine and Chlorine Dioxide (Amperometric), Ricca Chemical
For APHA Testing
$175.53 - $726.78
Specifications
| CAS | 7732-18-5,7558-79-4,7778-77-0 |
|---|---|
| For Use With (Application) | APHA for Chlorine and Chlorine Dioxide (Amperometric Method) |
| pH | 7 |
| Physical Form | Solution |
| Packaging | Natural Poly Bottle |
Specifications
| 7732-18-5,7558-79-4,7778-77-0 | |
| 7 | |
| Natural Poly Bottle |
| APHA for Chlorine and Chlorine Dioxide (Amperometric Method) | |
| Solution |