missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution for Nitrogen (Nitrate), Certified, LabChem™

Catalog No. LC125762
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC125762 1 L
LC125761 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC125762 Supplier LabChem Supplier No. LC125762

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Color Clear to slightly turbid
pH Range 3.0 ±0.5
Format Liquid
Packaging Amber Glass
Quantity 1 L
Storage Requirements Room Temperature
Type Buffer Solution for Nitrogen (Nitrate)