missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution for Nitrogen (Nitrate), Certified, LabChem™
Specifications
Specifications
| Color | Clear to slightly turbid |
| pH Range | 3.0 ±0.5 |
| Format | Liquid |
| Packaging | Amber Glass |
| Quantity | 500 mL |
| Storage Requirements | Room Temperature |
| Type | Buffer Solution for Nitrogen (Nitrate) |