missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, Giordano, pH 6.4 (M/15), for Wright Staining, Ricca Chemical

Catalog No. RC145016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RC145016 500 mL
145032 1 L
RC14501 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RC145016 Supplier Ricca Chemical Company Supplier No. RI145016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Wright Staining

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5, 7778-77-0, 7558-79-4
Color Colorless
For Use With (Application) Wright Staining
Physical Form Solution
Quantity 500 mL
Packaging Natural Poly Bottle