missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, Acetate, pH 4.0, For Residual Chlorine, APHA, Spectrum™ Chemical
Specifications
Specifications
| CAS | 6131-90-4 |
| pH Range | 3.5 to 4.2 |
| Purity or Quality Grade | Chemical Solution |
| Quantity | 4 L |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.