missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution A, for Sulfate, Certified, LabChem™

Catalog No. LC125702
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC125702 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC125702 Supplier LabChem Supplier No. LC125702
Only null left

Specifications

Chemical Name or Material Buffer Solution A, for Sulfate
Color Colorless
CAS Max % 94.4, 3, 2, 0.5, 0.1
CAS 7791-18-6, 64-19-7, 6131-90-4, 7757-79-1
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
pH 3.5 ±0.5
Physical Form Liquid
Quantity 1 L
Identification Passes Test
Linear Formula MgCl2·6H2O
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 203.31
Grade Certified
Show More Show Less