missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 13.0 (20°C), Glycine / sodium hydroxide / sodium chloride solution, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| pH | 13.0 |
| Quantity | 1 L |
| UN Number | NONH for all modes of transport |
| Chemical Name or Material | Buffer solution |