missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 3.56 ± 0.01 at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company 14985

Catalog No. 1498-5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Buffer, Reference Standard
877-24-7,7647-01-0,7732-18-5
100.0,1.04,0.06
20 L
Non-sterile
pH 3.56± 0.01 at 25°C
97.0,1.0,0.06
3.56
Cubitainer
Reference Standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions