missing translation for 'onlineSavingsMsg'
Learn More

Buffer pH 1.68, 500 mL, Mettler Toledo™

Catalog No. 01914028
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01914028 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01914028 Supplier METTLER TOLEDO Supplier No. 30258811
Only null left

NIST Traceable. Quality solutions for accurate results.

  • Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions.
  • Each standard solution comes with a certificate of analysis, providing traceability to NIST standards.

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Colorless
Description pH Buffer
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 1.68
Physical Form Liquid
Quantity 500 mL
Solution Type Calibration