missing translation for 'onlineSavingsMsg'
Learn More

Buffer concentrate, pH 13.00, glycine / sodium hydroxide / sodium chloride solution, Honeywell Fluka™

Catalog No. 6000384
Change view
Click to view available options
Quantity:
1 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000384 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000384 Supplier Honeywell Chemicals Supplier No. 387521EA
Only null left

for 500 ml buffer solution, glycine / sodium hydroxide / sodium chloride solution

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications

Chemical Name or Material Buffer concentrate
pH 13
Quantity 1 Pc.