missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Budesonide 98.0+%, TCI America™
$109.98 - $372.81
Chemical Identifiers
| CAS | 51333-22-3 |
|---|---|
| Molecular Formula | C25H34O6 |
| Molecular Weight (g/mol) | 430.54 |
| MDL Number | MFCD00083259,MFCD00138154 |
| InChI Key | VOVIALXJUBGFJZ-VXKMTNQYSA-N |
| Synonym | (+)-16alpha,17alpha-Butylidenedioxy-11beta,21-dihydroxy-1,4-pregnadiene-3,20-dione |
| PubChem CID | 73759663 |
| IUPAC Name | (1S,2S,4R,6R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.0²,â¹.0â´,â¸.0¹³,¹â¸]icosa-14,17-dien-16-one |
| SMILES | CCC[C@@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B3909200MG
|
TCI America
B3909200MG |
200 mg |
Each for $109.98
|
|
|||||
|
B39091G
|
TCI America
B39091G |
1 g |
Each for $372.81
|
|
|||||
Chemical Identifiers
| 51333-22-3 | |
| 430.54 | |
| VOVIALXJUBGFJZ-VXKMTNQYSA-N | |
| 73759663 | |
| CCC[C@@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO |
| C25H34O6 | |
| MFCD00083259,MFCD00138154 | |
| (+)-16alpha,17alpha-Butylidenedioxy-11beta,21-dihydroxy-1,4-pregnadiene-3,20-dione | |
| (1S,2S,4R,6R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.0²,â¹.0â´,â¸.0¹³,¹â¸]icosa-14,17-dien-16-one |
Specifications
| 51333-22-3 | |
| C25H34O6 | |
| 200 mg | |
| VOVIALXJUBGFJZ-VXKMTNQYSA-N | |
| (1S,2S,4R,6R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.0²,â¹.0â´,â¸.0¹³,¹â¸]icosa-14,17-dien-16-one | |
| 73759663 | |
| ≥98.0% (HPLC) | |
| Budesonide |
| White-Yellow | |
| MFCD00083259,MFCD00138154 | |
| (+)-16alpha,17alpha-Butylidenedioxy-11beta,21-dihydroxy-1,4-pregnadiene-3,20-dione | |
| CCC[C@@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO | |
| 430.54 | |
| 430.54 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : TU3723000
TSCA : No