Learn More
Thermo Scientific™ Buckets or Lids for Thermo Scientific™ TX-150 Swinging Bucket Rotor
Catalog No. 75005707
Change view
Click to view available options
Product Type:
Bucket
Bucket Lid
Capacity:
Not Applicable
8 x 50 mL
4 x 145 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Capacity
75005707 Bucket Lid Not Applicable
75005702 Bucket 4 x 145 mL
75005703 Bucket 8 x 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 75005707 Supplier Thermo Scientific™ Supplier No. 75005707
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Round and conical buckets offered in sets of four
Round and conical buckets or lids offered in sets of four
Specifications
For Use With (Equipment) Round Buckets
Capacity Not Applicable
Product Type Bucket Lid
Quantity Set of 4
Description ClickSeal Biocontainment Lids for Round Buckets