Learn More
Thermo Scientific™ Buckets or Lids for Thermo Scientific™ TX-150 Swinging Bucket Rotor
Catalog No. 75005702
Change view
Click to view available options
Type:
Bucket
Bucket Lid
Capacity:
Not Applicable
8 x 50 mL
4 x 145 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Capacity
75005702 Bucket 4 x 145 mL
75005707 Bucket Lid Not Applicable
75005703 Bucket 8 x 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 75005702 Supplier Thermo Scientific™ Supplier No. 75005702
Round and conical buckets offered in sets of four
Round and conical buckets or lids offered in sets of four
Specifications
Capacity 4 x 145 mL
Type Bucket
Quantity Set of 4
Description Round Buckets