missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Irradiated BucketLiners™

Catalog No. 19166564
Change view
Click to view available options
Packaging Type:
Bulk Pack, 6 Ea./Double Bag
Individually Double bagged
Dimensions:
45 x 45 cm (18 x 18 in.)
79 x 48 cm (31 x 19 in.)
89 x 53 cm; non-gusseted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type Dimensions
19166564 Individually Double bagged -
19305230 Individually Double bagged 45 x 45 cm (18 x 18 in.)
191304833 Individually Double bagged 79 x 48 cm (31 x 19 in.)
19116706 Individually Double bagged 89 x 53 cm; non-gusseted
19166902 Bulk Pack, 6 Ea./Double Bag -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19166564 Supplier Micronova™ Supplier No. BKL2430IR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Creates an impermeable barrier to strong solutions and raw materials when used to line cleanroom buckets and storage containers.

  • The unique LDPE/ Nylon laminate is a superb barrier material for lining totes to transport raw materials or to line buckets containing disinfectants or other cleaning solutions
  • Flat packed
  • Hold up to autoclave with liquid contents
  • Specifications

    Capacity (Metric) 22 L
    Material LDPE
    Autoclavable Not Autoclavable
    Disposable Single-use
    Length (English) 24 in.
    Packaging Type Individually Double bagged
    Sterilization Method Gamma-Irradiation