missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mopec Bucket Holder, ADA (HO009)
Description
Construction
- Designed to hold BA005 bucket on the exterior of the cart
- Will not interfere with hoods or ADA requirements
- Includes a locking pin
Materials
- 304 stainless steel construction
- 1 in. SQ. x 11 GA tubing
Dimensions
- 2.5 (arm-in) to 9.2 W (arm out) x 9.1 H in.
Performance
- Is removable (bolt-on)
- Can swivel when not in use
Compatibility
- BA005
- HA230-ADA
- HB1550
Specifications
Specifications
| Product Type | Bucket Holder |
| Height (English) | 9.1 in. |
| Width (English) | 2.5 to 9.2 in. |