missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Bucket and Cylinder (Archimedes' Principle)

Catalog No. S15903
Change view
Click to view available options
For Use With (Application):
Archimedes' Principle Apparatus
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S15903 Archimedes' Principle Apparatus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15903 Supplier United Scientific™ Supplier No. BCL001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for classroom use.

  • This Archimedes' Principal apparatus consists of a solid metal cylinder in a close-fitting metal bucket.
  • The bucket is 50 mm long and 19 mm in diameter
  • Displacement capacity approximately 22 mL
  • Shipping weight 3 oz.
  • Dimensions: 4 x 1.5 x 1.5 in.

Specifications

For Use With (Application) Archimedes' Principle Apparatus
Product Type Density Demonstration Kit

Warning: Cancer and reproductive harm