Learn More
BUCHI Glass U-Tube for Rotavapor R-220 condenser
Description
- Replacement OEM condenser piece
- Used between the expansion chamber and condenser
- Designed for Rotavapor R-220
Specifications
Specifications
| Material | Glass |
| Type | U-tube |
| For Use With (Equipment) | Rotavapor R-220 SE |
| Material (Coating) | Plastic |